L-Lysine 98%

Supplier: Thermo Scientific Chemicals

J62225-14 J62225-22 J62225-36
CAAAJ62225-14EA 201.53 CAD
CAAAJ62225-14 CAAAJ62225-22 CAAAJ62225-36
L-Lysine 98%
L-Lysine

Powder


Purity: 98% CAS No.: 56-87-1 Molecular Formula: C6H14N2O2 Formula Weight: 146.19 Physical Form: Powder Appearance: White Melting Point: 215° Solubility: Soluble in water Beilstein Reference: 1722531 Merck Reference: 14,5636 Optical Rotation: +26° (c=2 in 6M HCl) MDL No.: MFCD00064433

Formula: C₆H₁₄N₂O₂
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00064433
CAS Number: 56-87-1
EINECS: 200-294-2
Merck Index: 14,5636

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR