3,3'-Diaminobenzidine tetrahydrochloride hydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DAB HCl hydrate, 3,3',4,4'-Biphenyltetramine tetrahydrochloride hydrate, DAB tetrahydrochloride hydrate

J62216-06 J62216-09
CAAAJ62216-06EA 240.52 CAD
CAAAJ62216-06 CAAAJ62216-09
3,3'-Diaminobenzidine tetrahydrochloride hydrate 97%
3,3'-Diaminobenzidine tetrahydrochloride hydrate

Widely used peroxidase substrate for immunoblotting and immunohistochemical staining techniques.


Powder


Widely used peroxidase substrate for immunoblotting and immunohistochemical staining techniques

Formula: C₁₂H₁₄N₄·4HCl·H₂O
MW: 360.11 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD08273058
CAS Number: 868272-85-9
EINECS: 231-018-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR