L(+)-Serine cell culture reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine
J62187-09
J62187-22
J62187-36
J62187-A1
CAAAJ62187-09EA
75.32
CAD
CAAAJ62187-09
CAAAJ62187-22
CAAAJ62187-36
CAAAJ62187-A1
L(+)-Serine cell culture reagent
L(+)-Serine
Powder
CAS No.: 56-45-1
Molecular Formula: C3H7NO3
Formula Weight: 105.09
Physical Form: Powder
Appearance: White
Melting Point: 222°
Beilstein Reference: 1721404
Merck Reference: 14,8460
MDL No.: MFCD00064224
Formula:
C₃H₇NO₃ MW: 105.09 g/mol Boiling Pt: 259 Melting Pt: 222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064224 CAS Number: 56-45-1 EINECS: 200-274-3 Merck Index: 14,8460 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...