DL-Aminoglutethimide 99%

Supplier: Thermo Scientific Chemicals
Warning

J62182-03 J62182-06 J62182-14
CAAAJ62182-03EA 108.07 CAD
CAAAJ62182-03 CAAAJ62182-06 CAAAJ62182-14
DL-Aminoglutethimide 99%
DL-Aminoglutethimide

Adrenocortical suppressant which also inhibits conversion of androgens to estrogens by the aromatase enzyme system


Powder


Applications: Adrenocortical suppressant which also inhibits conversion of androgens to estrogens by the aromatase enzyme system

Formula: C₁₃H₁₆N₂O₂
Melting Pt: 152…154 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010122
CAS Number: 125-84-8
EINECS: 204-756-4
Merck Index: 14,440

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR