5-Aminofluorescein (isomer I) 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one (isomer I), 5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-Aminofluorescein, Fluoresceinamine, isomer I, 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, Fluoresceinamine (isomer I)

J62175-06 J62175-03
CAAAJ62175-06EA 1001.48 CAD
CAAAJ62175-06 CAAAJ62175-03
5-Aminofluorescein (isomer I) 96%
5-Aminofluorescein (isomer I)

A fluorescent labeling reagent


Powder


Applications: A fluorescent labeling reagent

Formula: C₂₀H₁₃NO₅
MW: 347.33 g/mol
Melting Pt: 223 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005052
CAS Number: 3326-34-9
EINECS: 222-043-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR