Adenosine-3',5'-cyclic monophosphate sodium salt 99%
Supplier: Thermo Scientific Chemicals
J62174-03
J62174-06
J62174-MD
CAAAJ62174-03EA
747.1
CAD
CAAAJ62174-03
CAAAJ62174-06
CAAAJ62174-MD
Adenosine-3',5'-cyclic monophosphate sodium salt 99%
Crystalline powder
Purity: 99%
CAS No.: 37839-81-9
Molecular Formula: C10H11N5NaO6P
Formula Weight: 351.19
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline powder
Appearance: White
Beilstein Reference: 54612
MDL No.: MFCD00069736
Formula:
C₁₀H₁₁N₅NaO₆P Storage Temperature: Freezer |
MDL Number:
MFCD00069736 CAS Number: 37839-81-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...