L(-)-Glutathione (reduced form) 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

J62166-06 J62166-14 J62166-22
CAAAJ62166-06EA 118.61 CAD
CAAAJ62166-06 CAAAJ62166-14 CAAAJ62166-22
L(-)-Glutathione (reduced form) 98+%
L(-)-Glutathione (reduced form)

A tripeptide used for reductive cleavage of disulfide bonds


Crystalline powder


A tripeptide used for reductive cleavage of disulfide bonds

Formula: C₁₀H₁₇N₃O₆S
MW: 307.33 g/mol
Melting Pt: ∼ 193 °C (dec.)
Density: 1.475 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 14,4475

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR