Amlodipine besylate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

J62164-03 J62164-06 J62164-14
CAAAJ62164-03EA 317.98 CAD
CAAAJ62164-03 CAAAJ62164-06 CAAAJ62164-14
Amlodipine besylate ≥98%
Amlodipine besylate

An L-type calcium channel antagonist with potent antioxidant activity


Solid


Applications: An L-type calcium channel antagonist with potent antioxidant activity

Formula: C₂₀H₂₅ClN₂O₅
Melting Pt: 195…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00887594
CAS Number: 111470-99-6
Merck Index: 14,491

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR