Zinc chloride 25 mM in aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

J62160-AD J62160-AE
CAAAJ62160-ADEA 97.04 CAD
CAAAJ62160-AD CAAAJ62160-AE
Zinc chloride 25 mM in aqueous solution
Zinc chloride

Liquid


CAS No.: 7646-85-7 Molecular Formula: Cl2Zn Formula Weight: 136.28 Physical Form: Liquid MDL No.: MFCD00011295

Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732
Melting Pt: 283
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
Merck Index: 13,10185

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR