Agarose Gel Loading Dye 6X for Electrophoresis, Alkaline, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62157-AB J62157-AC
CAAAJ62157-ABEA 175.14 CAD
CAAAJ62157-AB CAAAJ62157-AC
Agarose Gel Loading Dye 6X for Electrophoresis, Alkaline, Thermo Scientific
Stains Electrophoresis Stains
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.
Contains 300 mM sodium hydroxide, 6 mM EDTA, 0.25% bromocresol green, 0.25% xylene cyanol FF and 18% Ficoll
Order Now
SPECIFICATIONS
Environmentally Preferable