Sodium Phosphate, 0.2 M Buffer Solution, pH 7.4, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62152-AP
J62152-K2
CAAAJ62152-APEA
175.14
CAD
CAAAJ62152-AP
CAAAJ62152-K2
Sodium Phosphate, 0.2 M Buffer Solution, pH 7.4, Thermo Scientific
Buffers
Biological Buffers
Liquid
Physical Form: Liquid
MDL No.: MFCD00146206
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...