RBC lysis buffer for mouse, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62150-AK J62150-AP
CAAAJ62150-AKEA 121.41 CAD
CAAAJ62150-AK CAAAJ62150-AP
RBC lysis buffer for mouse, Thermo Scientific
Buffers Extraction Buffers

Liquid


Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Applications: For lysis of mouse red blood cells

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR