Disodium-β-glycerophosphate 200 mM in solution
Supplier: Thermo Scientific Chemicals
J62121-AD
J62121-AE
CAAAJ62121-ADEA
121.41
CAD
CAAAJ62121-AD
CAAAJ62121-AE
Disodium-β-glycerophosphate 200 mM in solution
Disodium β-glycerophosphate
Liquid
CAS No.: 819-83-0
Molecular Formula: C3H7Na2O6P
Formula Weight: 216.04
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00002135
Formula:
C₃H₇Na₂O₆P Storage Temperature: Ambient |
MDL Number:
MFCD00002135 CAS Number: 819-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...