Dextran sulfate sodium salt, M.W. ∼ 8,000
Supplier: Thermo Scientific Chemicals
J62101-14
J62101-22
CAAAJ62101-14EA
198.15
CAD
CAAAJ62101-14
CAAAJ62101-22
Dextran sulfate sodium salt, M.W. ∼ 8,000
Dextran sulphate sodium salt
Powder
CAS No.: 9011-18-1
Physical Form: Powder
Appearance: Off-white to pale yellow
Odor: Phenol-like
MDL No.: MFCD00081551
Formula:
(C₆H₇Na₃O₁₄S₃)n Storage Temperature: Ambient |
MDL Number:
MFCD00081551 CAS Number: 9011-18-1 Merck Index: 14,2951 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...