1-(1-Naphthyl)piperazine hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
J62097-03
J62097-06
CAAAJ62097-03EA
778.24
CAD
CAAAJ62097-03
CAAAJ62097-06
1-(1-Naphthyl)piperazine hydrochloride
1-(1-Naphthyl)piperazine hydrochloride
5-HT1 serotonin receptor agonist. 5-HT2 serotonin receptor antagonist
Solid
Applications: 5-HT1 serotonin receptor agonist. 5-HT2 serotonin receptor antagonist
Formula:
C₁₄H₁₆N₂·HCl MW: 248.75 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00055161 CAS Number: 104113-71-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...