ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) 95%
Supplier: Thermo Scientific Chemicals
J62089-MC
J62089-ME
CAAAJ62089-MCEA
373.59
CAD
CAAAJ62089-MC
CAAAJ62089-ME
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) 95%
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
Powder
Purity: 95%
CAS No.: 2646-71-1
Molecular Formula: C21H26N7Na4O17P3.3H2O
Formula Weight: 833.35
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to off-white
MDL No.: MFCD00036263
Formula:
C₂₁H₂₆N₇Na₄O₁₇P₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00036263 CAS Number: 2646-71-1 EINECS: 220-163-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...