Ofloxacin
Supplier: Thermo Scientific Chemicals
J62080-03
J62080-06
CAAAJ62080-03EA
93.95
CAD
CAAAJ62080-03
CAAAJ62080-06
Ofloxacin
Ofloxacin
Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity
Crystalline powder
Applications: Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity
Formula:
C₁₈H₂₀FN₃O₄ Melting Pt: 270…275 °C Storage Temperature: Freezer |
MDL Number:
MFCD00226105 CAS Number: 82419-36-1 Merck Index: 14,6771 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...