Maleate, 0.2 M Buffer Solution, pH 5.5, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62049-AK J62049-AP
CAAAJ62049-AKEA 120.82 CAD
CAAAJ62049-AK CAAAJ62049-AP
Maleate, 0.2 M Buffer Solution, pH 5.5, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 3105-55-3
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00012466

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR