Phosphate-Buffered Saline (PBS, 10X), pH 7.4, Thermo Scientific

Supplier: Thermo Scientific
J62036-K3 J62036-K2 J62036-K7 J62036-K7 J62036-K3 J62036-K2
CAAAJ62036-K7EA 323.81 CAD
AAJ62036-K2 CAAAJ62036-K7 CAAAJ62036-K2 CAAAJ62036-K3 AAJ62036-K7 AAJ62036-K3
Phosphate-Buffered Saline (PBS, 10X), pH 7.4, Thermo Scientific
Buffers Biological Buffers
Liquid

Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00131855
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR