Glimepiride

Supplier: Thermo Scientific Chemicals

J62032-03 J62032-MC
CAAAJ62032-03EA 1073.77 CAD
CAAAJ62032-03 CAAAJ62032-MC
Glimepiride
Glimepiride

A sulfonylurea hypoglecemic agent and potent potassium channel blocker


Powder


Applications: A sulfonylurea hypoglecemic agent and potent potassium channel blocker

Formula: C₂₄H₃₄N₄O₅S
Melting Pt: 212…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00878417
CAS Number: 93479-97-1
Merck Index: 14,4440

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR