Phosphate, 0.5 M Buffer Solution, pH 9.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62025-AK
J62025-AP
CAAAJ62025-AKEA
100.82
CAD
CAAAJ62025-AK
CAAAJ62025-AP
Phosphate, 0.5 M Buffer Solution, pH 9.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...