Fluconazole 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol, 2-(2,4-Difluorophenyl)-1,3-di(1H-1,2,4-triazol-1-yl)propan-2-ol

J62015-03 J62015-06 J62015-14
CAAAJ62015-03EA 303.06 CAD
CAAAJ62015-03 CAAAJ62015-06 CAAAJ62015-14
Fluconazole 99%
Fluconazole

Crystalline powder


Purity: 99% CAS No.: 86386-73-4 Molecular Formula: C13H12F2N6O Formula Weight: 306.27 Physical Form: Crystalline powder Density: 1.05 Appearance: White to off-white Melting Point: 138-140° MDL No.: MFCD00274549

Formula: C₁₃H₁₂F₂N₆O
MW: 306.28 g/mol
Melting Pt: 138…140 °C
Density: 1.05
Storage Temperature: Ambient
MDL Number: MFCD00274549
CAS Number: 86386-73-4
Merck Index: 14,4122

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR