Bisacodyl ≥98%

Supplier: Thermo Scientific Chemicals

J61997-09 J61997-14
CAAAJ61997-09EA 341.93 CAD
CAAAJ61997-09 CAAAJ61997-14
Bisacodyl ≥98%
Bisacodyl

A diphenolic laxative that stimulates adenyl cyclase activity


Crystalline powder


Applications: A diphenolic laxative that stimulates adenyl cyclase activity

Formula: C₂₂H₁₉NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00038039
CAS Number: 603-50-9
EINECS: 210-044-4
Merck Index: 14,1242

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR