Paraformaldehyde fixative solution

Supplier: Thermo Scientific Chemicals
Danger

J61984-AK J61984-AP
CAAAJ61984-AKEA 172.58 CAD
CAAAJ61984-AK CAAAJ61984-AP
Paraformaldehyde fixative solution
Paraformaldehyde

Liquid


Contains 3.7 % (w/v) paraformaldehyde and 3 % (w/v) sucrose in PBS, pH 7.4.

Formula: (CH₂O)n
Storage Temperature: Refrigerator
MDL Number: MFCD00133991
CAS Number: 30525-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR