MES-buffered saline (5X), pH 6.5, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61979-AK J61979-AP
CAAAJ61979-AKEA 194.31 CAD
CAAAJ61979-AK CAAAJ61979-AP
MES-buffered saline (5X), pH 6.5, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 145224-94-8
Physical Form: Liquid
MDL No.: MFCD00283848

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR