trans-Zeatin 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 6-(4-Hydroxy-3-methyl-2-butenylamino)purine

J61972-MC J61972-MD J61972-MF
CAAAJ61972-MCEA 1036.34 CAD
CAAAJ61972-MC CAAAJ61972-MD CAAAJ61972-MF
trans-Zeatin 97%
trans-Zeatin

Plant growth hormone that stimulates cell division


Powder


Applications: Plant growth hormone that stimulates cell division

Formula: C₁₀H₁₃N₅O
Melting Pt: 208…210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00213654
CAS Number: 1637-39-4
Merck Index: 14,10117

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR