Inosine 5'-monophosphate disodium salt hydrate
Supplier: Thermo Scientific Chemicals
J61959-06
J61959-14
J61959-22
CAAAJ61959-06EA
91.8
CAD
CAAAJ61959-06
CAAAJ61959-14
CAAAJ61959-22
Inosine 5'-monophosphate disodium salt hydrate
Inosine 5'-monophosphate disodium salt hydrate
Crystalline powder
CAS No.: 4691-65-0
Molecular Formula: C10H11N4Na2O8P.xH2O
Formula Weight: 392.17(anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
MDL No.: MFCD00036201
Formula:
C₁₀H₁₁N₄Na₂O₈P·xH₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD00150372 CAS Number: 352195-40-5 EINECS: 225-146-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...