Benztropine mesylate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cogentin

J61954-03 J61954-MC
CAAAJ61954-03EA 43.77 CAD
CAAAJ61954-03 CAAAJ61954-MC
Benztropine mesylate
Benztropine mesylate

Muscarinic receptor antagonist


Solid


Applications: Muscarinic receptor antagonist

Formula: C₂₂H₂₉NO₄S
Melting Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00074784
CAS Number: 132-17-2
EINECS: 205-048-8
UN: 2811
ADR: "6.1",III
Merck Index: 14,1121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR