Carbenicillin disodium salt

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na

J61949-03 J61949-06 J61949-14
CAAAJ61949-03EA 183.71 CAD
CAAAJ61949-03 CAAAJ61949-06 CAAAJ61949-14
Carbenicillin disodium salt
Carbenicillin disodium salt

Water-soluble antibiotic effective against Gram-negative bacteria.


Powder


Water-soluble antibiotic effective against Gram-negative bacteria

Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6
EINECS: 225-360-8
Merck Index: 14,1792

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR