Phosphoenolpyruvic Acid Tris(cyclohexylammonium) Salt ≥99%

Supplier: Thermo Scientific Chemicals

Synonyms: PEP-3CHA

J61943-03 J61943-06 J61943-14
CAAAJ61943-03EA 108.04 CAD
CAAAJ61943-03 CAAAJ61943-06 CAAAJ61943-14
Phosphoenolpyruvic Acid Tris(cyclohexylammonium) Salt ≥99%
Phosphoenolpyruvic acid tris(cyclohexylammonium)salt

Metabolic intermediate in many kinase reactions


Powder


Applications: Metabolic intermediate in many kinase reactions

Formula: C₃H₅O₆P·3C₆H₁₄N
MW: 465.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004258
CAS Number: 35556-70-8
EINECS: 252-618-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR