RNA sample loading buffer, no ethidium bromide, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J61937-S0
J61937-XF
CAAAJ61937-S0EA
242.85
CAD
CAAAJ61937-S0
CAAAJ61937-XF
RNA sample loading buffer, no ethidium bromide, Thermo Scientific
Buffers
Electrophoresis Buffers
Loading Buffers
Liquid
Physical Form: Liquid
MDL No.: MFCD00213041
Applications: Formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...