Metoprolol Tartrate ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(Isopropylamino)-3-[4-(beta-methoxyethyl)phenoxy]-2-propanol (+)-Tartrate

J61920-06 J61920-14
CAAAJ61920-06EA 183.04 CAD
CAAAJ61920-06 CAAAJ61920-14
Metoprolol Tartrate ≥98%
Metoprolol tartrate

A beta-adrenergic blocker


Powder


Applications: A beta-adrenergic blocker

Formula: C₁₅H₂₅NO₃
Melting Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00056257
CAS Number: 56392-17-7
EINECS: 260-148-9
Merck Index: 14,6151

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR