Phosphate-Buffered Saline (DPBS, 10X), Dulbecco's formula, Thermo Scientific

Supplier: Thermo Scientific
J61917-AP J61917-K2 J61917-K3 J61917-K3 J61917-AP J61917-K2
CAAAJ61917-APEA 76.71 CAD
CAAAJ61917-AP AAJ61917-AP CAAAJ61917-K2 AAJ61917-K2 AAJ61917-K3 CAAAJ61917-K3
Phosphate-Buffered Saline (DPBS, 10X), Dulbecco's formula, Thermo Scientific
Buffers Cell Culture Buffers
Liquid

Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00131855
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR