Luzindole 97%

Supplier: Thermo Scientific Chemicals

J61915-LB0 J61915-MB
CAAAJ61915-LB0EA 306.22 CAD
CAAAJ61915-LB0 CAAAJ61915-MB
Luzindole 97%
Luzindole

A melatonin receptor antagonist


Powder


A melatonin receptor antagonist

Formula: C₁₉H₂₀N₂O
Melting Pt: 44…46 °C
Storage Temperature: Freezer
MDL Number: MFCD00672498
CAS Number: 117946-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR