Enoxacin, crystalline powder
Supplier: Thermo Scientific Chemicals
A quinolone antibacteral agent which acts on DNA gyrase
Enoxacin is an antibacterial agent used in the treatment of gastro enteritis including infectious diarrhea, respiratory tract infections, gonorrhea and urinary tract infections.
It is a bactericidal and its mode of action depends on blocking of bacterial DNA replication by binding itself to an enzyme called DNA gyrase.
Caution: Store in cool place. Keep the container tightly closed in a dry and well-ventilated place.
Formula:
C₁₅H₁₇FN₄O₃ Melting Pt: 220…224 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00133308 CAS Number: 74011-58-8 Merck Index: 14,3587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...