Mycophenolic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

J61905-03 J61905-09 J61905-06
CAAAJ61905-03EA 108.07 CAD
CAAAJ61905-03 CAAAJ61905-09 CAAAJ61905-06
Mycophenolic acid 98%
Mycophenolic acid

Immunosupressant agent. Blocks inosine monophosphate dehydrogenase in the guanosine monophosphate pathway


Powder


Immunosupressant agent. Blocks inosine monophosphate dehydrogenase in the guanosine monophosphate pathway

Formula: C₁₇H₂₀O₆
MW: 320.34 g/mol
Melting Pt: 139…141 °C
Storage Temperature: Freezer
MDL Number: MFCD00036814
CAS Number: 24280-93-1
EINECS: 246-119-3
UN: 3077
ADR: 9 III
Merck Index: 14,6327

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR