L-Methionine cell culture reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
J61904-18
J61904-22
J61904-36
CAAAJ61904-18EA
226.11
CAD
CAAAJ61904-18
CAAAJ61904-22
CAAAJ61904-36
L-Methionine cell culture reagent
L-Methionine
Powder
CAS No.: 63-68-3
Molecular Formula: C5H11NO2S
Formula Weight: 149.21
Physical Form: Powder
Appearance: White
Melting Point: 284° dec.
Beilstein Reference: 1722294
Merck Reference: 14,5975
MDL No.: MFCD00063097
Formula:
C₅H₁₁NO₂S MW: 149.21 g/mol Boiling Pt: 278 Melting Pt: 136 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00063097 CAS Number: 63-68-3 EINECS: 200-562-9 Merck Index: 14,5975 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...