Paraformaldehyde 4% in PBS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Polyoxymethylene

J61899-AK J61899-AP
CAAAJ61899-AKEA 472.86 CAD
CAAAJ61899-AK CAAAJ61899-AP
Paraformaldehyde 4% in PBS
Paraformaldehyde

Liquid


Storage Temperature: 2°C to 8°C Physical Form: Liquid MDL No.: MFCD00133991

Formula: (CH₂O)n
Melting Pt: 123
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00133991
CAS Number: 30525-89-4
Merck Index: 13,07096

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR