Orange G/Blue Loading Dye (6X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
J61877-AB
J61877-AC
CAAAJ61877-ABEA
175.14
CAD
CAAAJ61877-AB
CAAAJ61877-AC
Orange G/Blue Loading Dye (6X), Thermo Scientific
Stains
Electrophoresis Stains
Orange G/blue loading dye (6X) is used for loading DNA samples into gel electrophoresis wells and tracking migration during electrophoresis.
- Contains: 0.25% Orange-G, 0.25% bromophenol blue, 0.25% xylene cyanol FF, 15% Ficoll, 30 mM EDTA in autoclaved DI water
It is used in molecular biology protocols and in nucleic acid gel electrophoresis and blotting. It is also used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.