TRIS-glycine native sample buffer (4X), pH 8.6, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61864-AC J61864-AD
CAAAJ61864-ACEA 121.41 CAD
CAAAJ61864-AC CAAAJ61864-AD
TRIS-glycine native sample buffer (4X), pH 8.6, Thermo Scientific
Buffers Electrophoresis Buffers Loading Buffers

Liquid


Physical Form: Liquid
Appearance: Colorless to yellow
MDL No.: MFCD00236360
Applications: For preparation of protein samples for native gel electrophoresis

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR