Glycine, 0.2 M buffer soln., pH 2.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J61855-AK
J61855-AP
CAAAJ61855-AKEA
121.41
CAD
CAAAJ61855-AK
CAAAJ61855-AP
Glycine, 0.2 M buffer soln., pH 2.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 56-40-6
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00008131
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...