Sodium pyruvate cell culture reagent

Supplier: Thermo Scientific Chemicals

J61840-18 J61840-22
CAAAJ61840-18EA 308.97 CAD
CAAAJ61840-18 CAAAJ61840-22
Sodium pyruvate cell culture reagent
Sodium pyruvate

Powder


CAS No.: 113-24-6 Molecular Formula: C3H3NaO3 Formula Weight: 110.04 Physical Form: Powder Appearance: White to pale yellow Solubility: Freely soluble in water Beilstein Reference: 3568341 MDL No.: MFCD00002586

Formula: CH₃COCOONa
Melting Pt: 220…230 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR