Magnesium sulfate heptahydrate cell culture reagent

Supplier: Thermo Scientific Chemicals

J61839-36 J61839-A1
CAAAJ61839-36EA 150.68 CAD
CAAAJ61839-36 CAAAJ61839-A1
Magnesium sulfate heptahydrate cell culture reagent
Magnesium sulphate heptahydrate

Powder


CAS No.: 10034-99-8 Molecular Formula: MgO4S.7H2O Formula Weight: 246.48 (120.37anhy) Physical Form: Powder Density: 1.68 Appearance: White Odor: Odorless Merck Reference: 14,5691 MDL No.: MFCD00149785

Formula: MgSO₄·7H₂O
Melting Pt: 1124 °C
Density: 1.68
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 14,5691

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR