Tolnaftate

Supplier: Thermo Scientific Chemicals

Synonyms: O-2-Naphthyl Methyl(3-methylphenyl)carbamothioate, O-Naphthalen-2-yl methyl(m-tolyl)carbamothioate, Methyl(3-methylphenyl)carbamothioic Acid O-2-Naphthyl Ester

J61834-03 J61834-06 J61834-14
CAAAJ61834-03EA 143.97 CAD
CAAAJ61834-03 CAAAJ61834-06 CAAAJ61834-14
Tolnaftate
Tolnaftate

Antifungal agent


Powder


Applications: Antifungal agent

Formula: C₁₉H₁₇NOS
MW: 307.41 g/mol
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00056611
CAS Number: 2398-96-1
EINECS: 219-266-6
Merck Index: 14,9519

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR