HEPES hemisodium salt

Supplier: Thermo Scientific Chemicals

Synonyms: N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid hemisodium salt, N-2-Hydroxyethylpiperazine-N'-2-ethanesulphonic acid hemisodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid hemisodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid hemisodium salt

J61830-22 J61830-30 J61830-36
CAAAJ61830-22EA 213.6 CAD
CAAAJ61830-22 CAAAJ61830-30 CAAAJ61830-36
HEPES hemisodium salt
HEPES hemisodium salt

A zwitterionic buffer


Crystalline powder


Applications: A zwitterionic buffer

Formula: C₈H₁₇.₅N₂O₄S·½Na
MW: 249.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS Number: 103404-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR