Spectinomycin dihydrochloride pentahydrate cell culture reagent

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate

J61820-06 J61820-14
CAAAJ61820-06EA 248.66 CAD
CAAAJ61820-06 CAAAJ61820-14
Spectinomycin dihydrochloride pentahydrate cell culture reagent
Spectinomycin dihydrochloride pentahydrate

An aminocyclitol antibiotic that binds to the 30S ribosomal subunit of bacteria and interrupts protein synthesis


White powder


Soluble in water

Formula: C₁₄H₂₄N₂O₇·2HCl·5H₂O
MW: 495.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150886
CAS Number: 22189-32-8
EINECS: 244-554-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR