Minoxidil

Supplier: Thermo Scientific Chemicals
Warning

J61803-03 J61803-06 J61803-14
CAAAJ61803-03EA 143.97 CAD
CAAAJ61803-03 CAAAJ61803-06 CAAAJ61803-14
Minoxidil
Minoxidil

Stimulates ATP-activated potassium channels


Crystalline powder


Applications: Stimulates ATP-activated potassium channels

Formula: C₉H₁₅N₅O
Melting Pt: 248…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00063409
CAS Number: 38304-91-5
EINECS: 253-874-2
Merck Index: 14,6203

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR