Isoproterenol hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

J61788-14 J61788-06
CAAAJ61788-14EA 538.51 CAD
CAAAJ61788-14 CAAAJ61788-06
Isoproterenol hydrochloride 98%
Isoproterenol hydrochloride

Standard selective beta-adrenoceptor agonist


Crystalline powder


Applications: Standard selective beta-adrenoceptor agonist

Formula: C₁₁H₁₇NO₃
Melting Pt: 165…175 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012603
CAS Number: 51-30-9
EINECS: 200-089-8
Merck Index: 14,5217

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR