Acid Blue 9

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 2-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulfonate, FD and C Blue No. 1, Brilliant Blue FCF, Erioglaucin A, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulphonate, Alphazurine FG, Erioglaucine sodium salt, CI 42090, Acid blue 9, Food Blue No.1

J61780-06 J61780-14
CAAAJ61780-06EA 52.39 CAD
CAAAJ61780-06 CAAAJ61780-14
Acid Blue 9
Erioglaucine disodium salt

Powder


CAS No.: 3844-45-9 Molecular Formula: C37H34N2Na2O9S3 Formula Weight: 792.86 Physical Form: Powder Appearance: Purple Melting Point: 283° MDL No.: MFCD00012141

Formula: C₃₇H₃₄Na₂N₂O₉S₃
MW: 792.84 g/mol
Melting Pt: 283 °C
Storage Temperature: Ambient
MDL Number: MFCD00012141
CAS Number: 3844-45-9
EINECS: 223-339-8
Merck Index: 14,1373

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR