Sulindac

Supplier: Thermo Scientific Chemicals
Danger

J61772-06 J61772-14
CAAAJ61772-06EA 114.08 CAD
CAAAJ61772-06 CAAAJ61772-14
Sulindac
Sulindac

A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells


Powder


Applications: A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells

Formula: C₂₀H₁₇FO₃S
Melting Pt: 182…185 °C
Storage Temperature: Freezer
MDL Number: MFCD00599589
CAS Number: 38194-50-2
EINECS: 253-819-2
UN: 2811
ADR: "6.1",III
Merck Index: 14,8982

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR